CAS Number 8017-16-1
Linear Formula Hn+2PnO3n+1
EC Number 232-417-0
MDL number MFCD00084480
| Related Categories | Chemical Synthesis, Inorganic Acids, Synthetic Reagents More... |
| assay | ≥83% phosphate (as P2O5) basis |
| bp | 300 °C(lit.) |
| cation traces | As: ≤30 mg/kg |
| Cu: ≤1 mg/kg | |
| Fe: ≤20 mg/kg | |
| Pb: ≤1 mg/kg |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?