11730 Sigma-Aldrich

Barium carbonate

purum p.a., ≥98.5% (KT)

DOWNLOAD MSDS (PDF)
  • CAS Number 513-77-9

  • Linear Formula BaCO3

  • Molecular Weight 197.34

  •  Beilstein Registry Number 7045119

  •  EC Number 208-167-3

  •  MDL number MFCD00003448

  •  PubChem Substance ID 24847325

Properties

Related Categories Chemical Synthesis, Inorganic Bases, Synthetic Reagents More...
grade   purum p.a.
assay   ≥98.5% (KT)
anion traces   chloride (Cl-): ≤300 mg/kg
cation traces   Ca: ≤1000 mg/kg
  Cd: ≤50 mg/kg
  Co: ≤50 mg/kg
  Cu: ≤50 mg/kg
  Fe: ≤50 mg/kg
  K: ≤100 mg/kg
  Na: ≤2000 mg/kg
  Ni: ≤50 mg/kg
  Pb: ≤50 mg/kg
  Sr: ≤1.5%
  Zn: ≤50 mg/kg

Price and Availability

Customers Also Viewed

Barium carbonate

99.999% trace metals basis

Barium carbonate

99.98% trace metals basis

Barium carbonate

ACS reagent, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?