EMAIL THIS PAGE TO A FRIEND
12262 Sigma-Aldrich

Omeprazole

pharmaceutical secondary standard; traceable to USP, PhEur and BP

Synonym: 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-1H-benzimidazole, Antra, Losec

  • CAS Number 73590-58-6

  • Empirical Formula (Hill Notation) C17H19N3O3S

  • Molecular Weight 345.42

  •  MDL number MFCD00083192

Purchase

Properties

grade   certified reference material
  pharmaceutical secondary standard
InChI Key   SUBDBMMJDZJVOS-UHFFFAOYSA-N
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   neat
pharmacopeia traceability   traceable to BP 765
  traceable to PhEur O0150000
  traceable to USP 1478505
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
DD9087000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?