Synonym: Charcoal bone
EC Number 232-421-2
MDL number MFCD00217223
PubChem Substance ID 24850849
| Related Categories | Activated Charcoal, Adsorbents, Filter Aids and Drying Agents, Essential Chemicals, Research Essentials |
| form | powder |
| loss | ≤5% loss on drying, 120°C; 3h |
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?