CAS Number 20816-12-0
Linear Formula OsO4
Molecular Weight 254.23
MDL number MFCD00011150
PubChem Substance ID 24852535
Popular Documents: Specification Sheet (PDF)
| Related Categories | Chemical Synthesis, Essential Chemicals, Inorganic Salts, Solutions and Reagents, Osmium, Oxidizing Agents, |
| vapor pressure | 5.42 psi ( 20 °C) |
| 6.76 psi ( 55 °C) | |
| contains | tert-butyl hydroperoxide as stabilizer |
| concentration | 2.5 wt. % in tert-butanol |
| density | 1.0 g/mL at 20 °C |
| 0.811 g/mL at 25 °C | |
| storage temp. | 2-8°C |
1, 5, 25 mL in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?