Synonym: Periodic acid potassium salt, Potassium (meta)periodate
CAS Number 7790-21-8
Linear Formula KIO4
Molecular Weight 230.00
EC Number 232-196-0
MDL number MFCD00011407
PubChem Substance ID 24852610
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Chemical Synthesis, Essential Chemicals, Hypervalent Iodine, Inorganic Salts, Solutions and Reagents, |
| grade | ACS reagent |
| assay | 99.8% |
| 99.8-100.3% (ACS specification) | |
| impurities | ≤0.01% other halogens |
| mp | 582 °C(lit.) |
| cation traces | Mn: ≤1 ppm |
100, 500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?