220736 Sigma-Aldrich

Florisil®

100-200 mesh

DOWNLOAD MSDS (PDF)

Synonym: Activated magnesium silicate, Magnesium silicate

  • CAS Number 1343-88-0

  • Empirical Formula (Hill Notation) MgO3Si

  • Molecular Weight 100.39

  •  EC Number 215-681-1

  •  MDL number MFCD00078249

Properties

Related Categories Adsorbents, Filter Aids and Drying Agents, Essential Chemicals, Florisil, Research Essentials
activation temperature   650 °C
particle size   100-200 mesh
surface area   289 m2/g

Description

Application

An activated magnesium silicate used as an adsorbent in gas chromatography.

Packaging

1 kg in glass bottle

250 g in glass bottle

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Price and Availability

Customers Also Viewed

Florisil<SUP>®</SUP>

reagent grade, 60-100 mesh

Florisil<SUP>®</SUP>

<200 mesh, fine powder

Florisil<SUP>®</SUP>

100-200 mesh, fine powder

30-60 mesh, granular

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?