CAS Number 10034-96-5
Linear Formula MnSO4 · H2O
Molecular Weight 169.02
EC Number 232-089-9
MDL number MFCD00149159
PubChem Substance ID 24853225
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Buffers / Buffer Salts, Chemical Synthesis, Core Bioreagents, Essential Chemicals, |
| grade | ACS reagent |
| assay | ≥98% |
| impurities | MnO4- reducers, passes test |
| ≤0.01% insolubles | |
| loss | 10.0-12.0% loss on ignition |
| anion traces | chloride (Cl-): ≤0.005% |
| cation traces | Ca: ≤0.005% |
100, 500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?