Synonym: Cobaltous nitrate hexahydrate, Cobaltous nitrate, Nitric acid, cobalt(II) salt
CAS Number 10026-22-9
Linear Formula Co(NO3)2 · 6H2O
Molecular Weight 291.03
MDL number MFCD00149647
PubChem Substance ID 24854365
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, Catalysis and Inorganic Chemistry, Chemical Synthesis, Cobalt, Essential Chemicals, |
| grade | ACS reagent |
| assay | ≥98% |
| 98.0-102.0% | |
| impurities | ≤0.01% insolubles |
| mp | 55 °C(lit.) |
| anion traces | chloride (Cl-): ≤0.002% |
| sulfate (SO42-): ≤0.005% |
100, 500 g in poly bottle
5 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?