25044 Sigma-Aldrich

Zinc acetate dihydrate

puriss., meets analytical specification of DAB, E 650, 99-102%

DOWNLOAD MSDS (PDF)
  • CAS Number 5970-45-6

  • Linear Formula Zn(CH3COO)2 · 2H2O

  • Molecular Weight 219.51

  •  Beilstein Registry Number 3732513

  •  EC Number 209-170-2

  •  MDL number MFCD00066961

  •  PubChem Substance ID 24855059

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Zinc More...
grade   puriss.
assay   99-102%
quality   meets analytical specification of DAB, E 650
impurities   organic volatile impurities, complies (GC)
  ≤0.005% insoluble matter in CH3COOH
  ≤0.2% alkalis/earth alkalis
pH   6-8 (20 °C, 5%)
anion traces   chloride (Cl-): ≤50 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   As: ≤3 mg/kg
  Ca: ≤50 mg/kg
  Cd: ≤5 mg/kg
  Fe: ≤50 mg/kg
  K: ≤50 mg/kg
  Mg: ≤50 mg/kg
  Na: ≤50 mg/kg
  Pb: ≤20 mg/kg

Price and Availability

Customers Also Viewed

Zinc acetate

99.99% trace metals basis

Zinc acetate dihydrate

99.999% trace metals basis

Zinc acetate dihydrate

ACS reagent, ≥98%

Zinc acetate dihydrate

reagent grade

Zinc acetate dihydrate

puriss. p.a., ACS reagent, ≥99.0% (KT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?