255823 Sigma-Aldrich

Lithium carbonate

ACS reagent, ≥99.0%

DOWNLOAD MSDS (PDF)

Synonym: Carbolithium, Carbonic acid lithium salt

  • CAS Number 554-13-2

  • Linear Formula Li2CO3

  • Molecular Weight 73.89

  •  Beilstein Registry Number 3999191

  •  EC Number 209-062-5

  •  MDL number MFCD00011084

  •  PubChem Substance ID 24855485

Properties

Related Categories ACS Grade, Chemical Synthesis, Essential Chemicals, Inorganic Bases, Inorganic Salts, Solutions and Reagents,
grade   ACS reagent
assay   ≥99.0%
impurities   ≤0.01% insol. dil. HCl
  ≤0.2% S compounds
mp   618 °C(lit.)
anion traces   chloride (Cl-): ≤0.005%
  nitrate (NO3-): ≤5 ppm
cation traces   Ca: ≤0.01%
  Fe: ≤0.002%
  K: ≤0.01%
  NH4+: ≤5 ppm
  Na: ≤0.1%
  heavy metals (as Pb): ≤0.002%

Description

Packaging

100, 500 g in poly bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Lithium carbonate

TraceSELECT®, for trace analysis, ≥99.998% (metals basis)

Lithium carbonate

puriss. p.a., ACS reagent, reagent (for microscopy), ≥99.0% (T)

Lithium carbonate

purum p.a., ≥98.0% (T)

Lithium carbonate

purum, ≥99%

Lithium carbonate

99.997% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?