CAS Number 120-82-1
Empirical Formula (Hill Notation) C6H3Cl3
Molecular Weight 181.45
Beilstein Registry Number 956819
EC Number 204-428-0
MDL number MFCD00000547
PubChem Substance ID 24855525
Popular Documents: Specification Sheet (PDF)
| Related Categories | Amber Glass Bottles, Solvent Bottles, Solvent Packaging Options, Solvent by Application, Solvents, |
| grade | CHROMASOLV® |
| vapor density | >6 (vs air) |
| vapor pressure | 1 mmHg ( 40 °C) |
| assay | ≥99% |
| autoignition temp. | 1060 °F |
| expl. lim. | 6.6 %, 150 °F |
| impurities | <0.020% water |
1, 2 L in glass bottle
4, 4×4 L in PVC coated
CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?