Synonym: PCA
CAS Number 7601-90-3
Linear Formula HClO4
Molecular Weight 100.46
MDL number MFCD00011325
PubChem Substance ID 24858522
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Acids & Bases, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, |
| grade | ACS reagent |
| vapor pressure | 7 mmHg ( 25 °C) |
| assay | 60% |
| 60.0-62.0% (ACS specification) | |
| impurities | ≤0.001% N compounds |
| ≤5 ppm Silicate and phosphate (as SIO2) | |
| ign. residue | ≤0.003% |
ACS specifications same as product 244252, except for concentration
1, 2.5 L in glass bottle
100, 500 mL in glass bottle
Customers Also Viewed

puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?