Synonym: Ammonia aqueous, Ammonia water
CAS Number 1336-21-6
Linear Formula NH4OH
Molecular Weight 35.05
Beilstein Registry Number 3587154
MDL number MFCD00066650
PubChem Substance ID 24859252
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS Grade, ACS Grade Acids & Bases, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, |
| grade | ACS reagent |
| vapor density | 1.2 (vs air) |
| vapor pressure | 115 mmHg ( 20 °C) |
| assay | 28.0-30.0% NH3 basis |
| expl. lim. | 27 % |
| impurities | MnO4- reducers, passes test |
| ≤0.002% CO2 |
2.5, 6×2.5 L in PVC coated
500, 6×500 mL in PVC coated
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?