412236 Sigma-Aldrich

Octane

reagent grade, 98%

DOWNLOAD MSDS (PDF)

Synonym: n-Octane

  • CAS Number 111-65-9

  • Linear Formula CH3(CH2)6CH3

  • Molecular Weight 114.23

  •  Beilstein Registry Number 1696875

  •  EC Number 203-892-1

  •  MDL number MFCD00009556

  •  PubChem Substance ID 24865796

Properties

Related Categories ACS and Reagent Grade Solvents, Amber Glass Bottles, Reagent, Reagent Grade Solvents, Solvent Bottles,
grade   reagent grade
vapor density   3.9 (vs air)
vapor pressure   11 mmHg ( 20 °C)
assay   98%
autoignition temp.   428 °F
expl. lim.   6.5 %
refractive index   n20/D 1.398(lit.)
bp   125-127 °C(lit.)
mp   −57 °C(lit.)
density   0.703 g/mL at 25 °C(lit.)

Description

Packaging

1 L in glass bottle

100, 500 mL in glass bottle

Price and Availability

Customers Also Viewed

Decane
D901Decane

ReagentPlus®, ≥99%

Octane
296988Octane

anhydrous, ≥99%

Octane
657042Octane

electronic grade, ≥99.999% metals basis, ≥99% (CP)

Octane
74820Octane

analytical standard

Octane
74821Octane

puriss. p.a., ≥99.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?