414220 Sigma-Aldrich

Propylene carbonate

CHROMASOLV®, for HPLC, 99.7%

DOWNLOAD MSDS (PDF)

Synonym: 1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one

  • CAS Number 108-32-7

  • Empirical Formula (Hill Notation) C4H6O3

  • Molecular Weight 102.09

  •  Beilstein Registry Number 107913

  •  EC Number 203-572-1

  •  MDL number MFCD00005385

  •  PubChem Substance ID 24865903

Properties

Related Categories Alternative Energy, Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, CHROMASOLV for HPLC,
grade   CHROMASOLV®
  for HPLC
vapor pressure   0.13 mmHg ( 20 °C)
  0.98 mmHg ( 50 °C)
assay   99.7%
autoignition temp.   851 °F
expl. lim.   14.3 %
impurities   <0.020% water
evapn. residue   <0.0005%
refractive index   n20/D 1.421(lit.)
bp   240 °C(lit.)
mp   −55 °C(lit.)
density   1.204 g/mL at 25 °C(lit.)
λ   H2O reference
UV absorption   λ: 235 nm Amax: 1.00
  λ: 280 nm Amax: 0.50
  λ: 300 nm Amax: 0.30
  λ: 350 nm Amax: 0.05
  λ: 375-400 nm Amax: 0.01

Description

Packaging

1, 2 L in glass bottle

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

Propylene carbonate

Selectophore, ≥99.0%

Propylene carbonate

anhydrous, 99.7%

Propylene carbonate

ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?