496189 Sigma-Aldrich

Chloroform

biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer

DOWNLOAD MSDS (PDF)

Synonym: Methylidyne trichloride, Trichloromethane

Properties

Related Categories Biotech, Biotech Solvents, Chloroform, Core Bioreagents, General Reagents,
grade   biotech. grade
vapor density   4.1 (vs air)
vapor pressure   160 mmHg ( 20 °C)
assay   ≥99.8%
contains   0.5-1.0% ethanol as stabilizer
impurities   <0.01% water
evapn. residue   <0.0003%
color   APHA: ≤10
refractive index   n20/D 1.445(lit.)
pH   5.2-6.4
bp   60.5-61.5 °C(lit.)
mp   −63 °C(lit.)
density   1.48 g/mL at 25 °C
  1.492 g/mL at 25 °C(lit.)
cation traces   Pb: ≤0.05 ppm

Description

Packaging

1, 2 L in Sure/Seal™

Price and Availability

Customers Also Viewed

2-Propanol

BioReagent, for molecular biology, ≥99%

Chloroform

contains 100-200 ppm amylenes as stabilizer, ≥99.5%

Chloroform

≥99%, PCR Reagent, contains amylenes as stabilizer

Chloroform

anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer

Chloroform

contains amylenes as stabilizer, ACS reagent, ≥99.8%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?