71657 Sigma-Aldrich

Sodium bisulfate monohydrate

puriss. p.a., ≥99.0% (T)

DOWNLOAD MSDS (PDF)

Synonym: Sodium hydrogen sulfate monohydrate

  • CAS Number 10034-88-5

  • Linear Formula NaHSO4 · H2O

  • Molecular Weight 138.08

  •  EC Number 231-665-7

  •  MDL number MFCD00149210

  •  PubChem Substance ID 24886244

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Puriss p.a., Q-S, Puriss p.a. More...
grade   puriss. p.a.
assay   ≥99.0% (T)
impurities   ≤0.001% total nitrogen (N)
anion traces   chloride (Cl-): ≤10 mg/kg
  phosphate (PO43-): ≤5 mg/kg
cation traces   Ag: ≤5 mg/kg
  Al: ≤5 mg/kg
  Ba: ≤5 mg/kg
  Bi: ≤5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤20 mg/kg
  Li: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  Mo: ≤5 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Sr: ≤5 mg/kg
  Tl: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Other Notes

Reagent for the decomposition of basic metal oxides by fusion1,2

Price and Availability

Customers Also Viewed

Sodium bisulfate

purum, anhydrous, ~95.0%

Sodium bisulfate

technical, anhydrous, ≥93%

Sodium bisulfate monohydrate

ReagentPlus®, 99%

Sodium bisulfite

ACS reagent, mixture of NaHSO3 and Na2S2O5

Sodium hydrogen sulfate

technical grade

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?