84422 Sigma-Aldrich

Hydrochloric acid

puriss. p.a., ACS reagent, fuming, ≥37% (T)

DOWNLOAD MSDS (PDF)
  • CAS Number 7647-01-0

  • Empirical Formula (Hill Notation) HCl

  • Molecular Weight 36.46

  •  Beilstein Registry Number 1098214

  •  EC Number 231-595-7

  •  MDL number MFCD00011324

Properties

Related Categories ACS Grade, ACS Grade Acids & Bases, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use,
grade   ACS reagent
vapor density   1.3 (vs air)
vapor pressure   3.23 psi ( 21.1 °C)
  7.93 psi ( 37.7 °C)
grade   puriss. p.a.
assay   ≥37% (T)
impurities   ≤0.00005% free chlorine (Cl)
ign. residue   ≤0.0005% (as SO4)
bp   >100 °C(lit.)
density   1.18 g/mL at 20 °C
  1.2 g/mL at 25 °C(lit.)
anion traces   bromide (Br-): ≤50 mg/kg
  phosphate (PO43-): ≤0.5 mg/kg
  sulfate (SO42-): ≤1 mg/kg
  sulfite (SO32-): ≤1 mg/kg
cation traces   Ag: ≤0.01 mg/kg
  Al: ≤0.1 mg/kg
  As: ≤0.01 mg/kg
  Ba: ≤0.02 mg/kg
  Bi: ≤0.1 mg/kg
  Ca: ≤0.5 mg/kg
  Cd: ≤0.01 mg/kg
  Co: ≤0.01 mg/kg
  Cr: ≤0.02 mg/kg
  Cu: ≤0.02 mg/kg
  Fe: ≤0.2 mg/kg
  Hg: ≤0.01 mg/kg
  K: ≤0.1 mg/kg
  Li: ≤0.01 mg/kg
  Mg: ≤0.1 mg/kg
  Mn: ≤0.01 mg/kg
  Mo: ≤0.02 mg/kg
  NH4+: ≤1 mg/kg
  Na: ≤0.5 mg/kg
  Ni: ≤0.02 mg/kg
  Pb: ≤0.02 mg/kg
  Sr: ≤0.01 mg/kg
  Tl: ≤0.05 mg/kg
  Zn: ≤0.05 mg/kg

Description

Other Notes

Sales restrictions may apply

Price and Availability

Customers Also Viewed

Hydrochloric acid

≥37%, TraceSELECT®, for trace analysis, fuming

Hydrochloric acid

ACS reagent, 37%

Hydrochloric acid

ACS reagent, 37%

Hydrochloric acid

puriss. p.a., ACS reagent, fuming 37%, packed in coated, shock- and leak-protected glass bottle, ≥37% (T)

Hydrochloric acid

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?