Synonym: Hartshorn salt
CAS Number 506-87-6
Linear Formula (NH4)2CO3
Molecular Weight 96.09
Beilstein Registry Number 4253487
EC Number 208-058-0
MDL number MFCD00010890
PubChem Substance ID 24891462
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Chemical Synthesis, Inorganic Bases, Synthetic Reagents More... |
| vapor density | 2.7 (vs air) |
| composition | ammonia, ≥30% |
May solidify on storage
1 kg in poly bottle
100, 500 g in poly bottle
A mixture of ammonium bicarbonate and ammonium carbamate.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?