Synonym: Charcoal activated
CAS Number 7440-44-0
Empirical Formula (Hill Notation) C
Molecular Weight 12.01
EC Number 231-153-3
MDL number MFCD00133992
PubChem Substance ID 24892524
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Activated Charcoal, Activated and Mesoporous Carbon, Adsorbents, Filter Aids and Drying Agents, Alternative Energy, Essential Chemicals, |
| vapor pressure | <0.1 mmHg ( 20 °C) |
| form | untreated, granular |
| autoignition temp. | 842 °F |
| resistivity | 1375 μΩ-cm20°C (graphite) |
| particle size | 4-8 mesh |
| mp | 3550 °C(lit.) |
2.5 kg in poly drum
500 g in poly bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?