• USA Home
  • F5645 - Fura 2 magnesium-selective analog tetrapotassium salt

EMAIL THIS PAGE TO A FRIEND
F5645 Sigma-Aldrich

Fura 2 magnesium-selective analog tetrapotassium salt

~90% (TLC)

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Ion Channels, Ion Probes, Monovalent Ion Channels,
InChI Key   HPJZOFRYSYPJGW-UHFFFAOYSA-N
assay   ~90% (TLC)
storage temp.   −20°C

Description

Application

Cell impermeable fluorescent probe for Mg2+

Price and Availability


All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?