Synonym: Isoamyl alcohol, Isopentyl alcohol
CAS Number 123-51-3
Linear Formula (CH3)2CHCH2CH2OH
Molecular Weight 88.15
Beilstein Registry Number 1718835
EC Number 204-633-5
MDL number MFCD00002934
PubChem Substance ID 24896825
Popular Documents: Specification Sheet (PDF)
| Related Categories | ACS and Reagent Grade Solvents, Amber Glass Bottles, Carbon Steel Flex-Spout Cans, Humulus lupulus (Hops), Nutrition Research, |
| grade | reagent grade |
| vapor density | 3 (vs air) |
| vapor pressure | 2 mmHg ( 20 °C) |
| assay | 98% |
| autoignition temp. | 644 °F |
| expl. lim. | 1.2-9 %, 100 °F |
| refractive index |
n |
1, 4 L in glass bottle
100, 500 mL in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?