Synonym: Caustic potash
CAS Number 1310-58-3
Linear Formula KOH
Molecular Weight 56.11
EC Number 215-181-3
MDL number MFCD00003553
PubChem Substance ID 24898721
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Acids & Bases, Core Bioreagents, Life Science Reagents for Cloning, Life Science Reagents for DNA/RNA Electrophoresis, Life Science Reagents for Protein Expression and Purification, |
| vapor pressure | 1 mmHg ( 719 °C) |
| product line | BioXtra |
| assay | ≥85% KOH basis |
| impurities | ≤0.0005% Phosphorus (P) |
| ≤0.1% Insoluble matter | |
| ≤2.0% K2CO3 | |
| mp | 361 °C(lit.) |
Customers Also Viewed

semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?