Synonym: ‘Caustic soda’
CAS Number 1310-73-2
Linear Formula NaOH
Molecular Weight 40.00
EC Number 215-185-5
MDL number MFCD00003548
PubChem Substance ID 24899676
Popular Documents: Product Information Sheet (PDF) | Specification Sheet (PDF)
| Related Categories | Alkaline β-elimination, Chemical Deglycosylation, Deglycosylation Strategies, Essential Chemicals, Glycobiology, |
| grade | reagent grade |
| vapor density | >1 (vs air) |
| vapor pressure | <18 mmHg ( 20 °C) |
| 3 mmHg ( 37 °C) | |
| assay | ≥98% |
| form | pellets (anhydrous) |
| impurities | ≤1.0% sodium carbonate |
1, 6×1 kg in glass bottle
Bulk packages
5 kg in poly drum
500 g in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?