| Related Categories | A-C, Cell Culture, ECACC Cell Lines More... |
| biological source | Ovarian metastasis from human |
| growth mode | Adherent |
| karyotype | Modal chromosome no. 38 (36-42) |
| morphology | Polygonal & fusiform |
| shipped in | dry ice |
| storage temp. | −196°C |
For complete product information, please see COV413B
DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)
Split confluent cultures 1:4 to 1:10 i.e. seeding at 2-4x10,000 cells/cm2 using trypsin/EDTA; 5% CO2; 37°C.
A human ovarian epithelial-serous carcinoma cell line established from an intraperitoneal metastasis of the bladder dome. This cell line was derived from the same patient as the ovarian cell line COV413A (ECACC cat no. 07071905). Anchorage-independent growth in agar has been reported.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?