84110801 Sigma

Myb2-7

Reactivity: Anti-myb gene products, avian

DOWNLOAD MSDS (PDF)

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridomas More...
biological source   from Mouse x Mouse Hybridoma
ImmunoDon   BALB/c mouse spleen
Myeloma   Sp2/0
isotype   IgG1, kappa
shipped in   dry ice
storage temp.   −196°C

Description

Immunogen

The bacterially expressed fusion protein, bp37v-myb

General description

For complete product information, please see Myb2-7

Culture Medium

RPMI 1640 + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

The antibody recognises an antigen on the primary structure of bp37v-myb and cross reacts with v-myb gene product of 45kDa and c-myb gene product of 75kDa. The antibody recognises E26 oncogene product p135gag-myb-ets.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?