85011423 Sigma

BHK TK-

DOWNLOAD MSDS (PDF)

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   kidney from hamster
growth mode   Adherent
morphology   Fibroblast
shipped in   dry ice
storage temp.   −196°C

Description

Application

Transformation and virus studies

General description

For complete product information, please see BHK TK-

Culture Medium

DMEM or GMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 i.e. seeding at 1-2x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Cell Line Description

BHK TK- is a derivative of BHK21 (clone 13) rendered TK- by selection in 30μg/ml BUdR. Used widely for research on viruses and on neoplastic transformation especially with polyoma virus.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?