85011441 Sigma

A2H

DOWNLOAD MSDS (PDF)

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   ovary from hamster
growth mode   Adherent
karyotype   2n = 22
morphology   Epithelial
shipped in   dry ice
storage temp.   −196°C

Description

Application

Genetic studies

General description

For complete product information, please see A2H

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Cell Line Description

Used for the induction and isolation of nutritionally deficient mutants. A reclone of ′Clone A′ established by Puck (the ′H′ denotes MRC Harwell). Useful for genetic studies due to the low chromosome number.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?