85042304 Sigma

B92

DOWNLOAD MSDS (PDF)

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   nerve from rat
growth mode   Adherent
morphology   Glial/neuronal
shipped in   dry ice
storage temp.   −196°C

Description

Application

Neurobiological studies

General description

For complete product information, please see B92

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Cell Line Description

Ethyl nitrosaurea induced tumour cell line. Neuronal and glial cells are useful for research into the distribution of neurotransmitter synthesis and brain specific antigens among nerve and glia. Morphology significantly affected when grown in serum free media.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?