85102267 Sigma

AFRC MAC 5

Reactivity: Anti-Glycoprotein of Chlamydomonas reinhardii

DOWNLOAD MSDS (PDF)

Properties

Related Categories Cell Culture, ECACC Cell Lines, Hybridomas More...
biological source   from Rat x Rat Hybridoma
ImmunoDon   AO rat spleen
Myeloma   YB2/3.0 Ag20
isotype   IgG2b
shipped in   dry ice
storage temp.   −196°C

Description

Immunogen

2BII, the major glycoprotein fraction of the cell wall of Chlamydomonas reinhardii

General description

For complete product information, please see AFRC MAC 5

Culture Medium

DMEM + 2mM glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. These cells require mouse macrophage feeder layers upon resuscitation

Cell Line Description

The antibody recognises the glycoprotein 2BII and labels both the whole cell surface and also the flagella. Binding is lost after acid hydrolysis or de-glycosylation of 2BII. AFRC MAC 5 antibody shows similar fine specificity and properties to AFRC MAC 3 and 4.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?