86093002 Sigma

CRFK (BVD Ag negative)

DOWNLOAD MSDS (PDF)

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   Kidney from feline
growth mode   Adherent
karyotype   Hypodiploid, modal no 36
morphology   Epithelial
products   Carcinoembryonic antigen (CEA)
shipped in   dry ice
storage temp.   −196°C

Description

Application

Virus studies

General description

For complete product information, please see CRFK (BVD Ag negative)

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:8 i.e. seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Cell Line Description

CRFK cells were isolated from the cortical portion of the kidneys of a 10-12 week old normal female domestic cat. CRFK are extensively used in feline virus research. Susceptible to herpesvirus and feline picornaviruses producing CPE. BVD Ag negative.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?