87022601 Sigma

BFA

DOWNLOAD MSDS (PDF)

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   aorta from bovine
growth mode   Adherent
morphology   Endothelial
products   Collagen type 3
shipped in   dry ice
storage temp.   −196°C

Description

Application

Virus studies

General description

For complete product information, please see BFA

Culture Medium

Ham′s F12 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Cell Line Description

Derived from a bovine foetus.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Customers Also Viewed

Brefeldin A

from Penicillium brefeldianum, ≥99% (HPLC and TLC)

Brefeldin A

≥99% (HPLC and TLC), BioXtra, for molecular biology

Brefeldin A

from Penicillium brefeldianum, Ready Made Solution, 10 mg/mL in DMSO, 0.2 μm filtered

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?