87061209 Sigma

COLO 668

DOWNLOAD MSDS (PDF)

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   Lung (brain metastasis) from human
growth mode   Suspension
karyotype   2n = 46
morphology   Round & Refractile
shipped in   dry ice
storage temp.   −196°C

Description

Application

Tumourigenicity studies

General description

For complete product information, please see COLO 668

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10
D13S317: 10,11
D16S539: 11,12
D5S818: 11
D7S820: 8.2,10
THO1: 9
TPOX: 8
vWA: 16

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Cell Line Description

Derived from a brain metastasis of a 47 year old female with oat cell carcinoma of the lung.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?