88102402 Sigma

AV3 (HeLa derivative)

DOWNLOAD MSDS (PDF)

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   Cervix from human
growth mode   Adherent
karyotype   2n = 46
morphology   Epithelial
shipped in   dry ice
storage temp.   −196°C

Description

Application

Virus studies: poliovirus 1

General description

For complete product information, please see AV3 (HeLa derivative)

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 9,10
D13S317: 14
D16S539: 9,10
D5S818: 11,12
D7S820: 12
THO1: 7
TPOX: 8,12
vWA: 16,18

Culture Medium

EMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 2-5x100,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Cell Line Description

This cell line was originally derived from normal human amnion. The cells are mainly used for virus studies (susceptible to poliovirus 1). HeLa marker chromosomes and type A G6PD have been detected. This cell line was found to be indistinguishable from HeLa by STR PCR DNA profiling. Therefore, the cell line must be considered to be derived from HeLa.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?