90031532 Sigma

BUD-8

DOWNLOAD MSDS (PDF)

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   skin from human
growth mode   Adherent
karyotype   Diploid
morphology   Fibroblast
shipped in   dry ice
storage temp.   −196°C

Description

Application

Interferon assay

General description

For complete product information, please see BUD-8

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 4x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C.

Cell Line Description

Derived from the normal skin of a 56 year old Caucasian female. The cells are highly sensitive to human interferon and have been proposed as a standard for its assay.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?