93040120 Sigma

BE10-Late

DOWNLOAD MSDS (PDF)

Properties

Related Categories A-C, Cell Culture, ECACC Cell Lines More...
biological source   brain from rat
growth mode   Adherent
products   Plasminogen activator activity
shipped in   dry ice
storage temp.   −196°C

Description

Application

Study of carcinogenesis, tumourigenicity studies

General description

For complete product information, please see BE10-Late

Culture Medium

DMEM + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:10 to 1:20 i.e. seeding at 1-2x100,000 cells/25cm2 using 0.25% trypsin/EDTA; 10-15% CO2; 37°C.

Cell Line Description

Malignant cells produced on passaging BE10-Intermediate (Sigma Catalogue number. 93040119) beyond passage 40. The cells possess tissue plasminogen activator activity, are tumourigenic in BDIX rats and form colonies in agar.

Price and Availability

NEW! Sigma® Cultureware


Cell Culture SuppliesIntroducing Sigma’s new competitively
priced line of cultureware:

  • . Multiwell Plates
  • . Culture Flasks
  • . Cryogenic Vials & More



Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?