| Related Categories | Cell Culture, ECACC Cell Lines, J-N More... |
| biological source | Skin from deer |
| growth mode | Adherent |
| karyotype | 2n = 62 |
| morphology | Fibroblast-like |
| shipped in | dry ice |
| storage temp. | −196°C |
For complete product information, please see KCB 89001
Medium 199 + 2mM Glutamine + 15% Foetal Bovine Serum (FBS).
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.
Derived from the skin of an adult, female Sambar deer (Cervus unicolor), in January 1989.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?