| Related Categories | A-C, Cell Culture, ECACC Cell Lines More... |
| biological source | Lung (bronchus) from human |
| growth mode | Adherent |
| morphology | Epithelial |
| shipped in | dry ice |
| storage temp. | −196°C |
For complete product information, please see ChaGo-K-1
RPMI 1640 + 2mM Glutamine + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.
This cell line was derived from a bronchogenic carcinoma of a 45-year-old male. The cells produce high levels of MUC-1 mucin mRNA, low levels of MUC-2 mRNA but do not express the MUC-3 gene.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?