AQ0125 Sigma

AQUA Peptide LVINGNPITIFQE[R]

DOWNLOAD MSDS (PDF)
  • Empirical Formula (Hill Notation) C7213C6H127N1715N4O22

  • Molecular Weight 1720.90

Properties

Related Categories AQUA Peptides, Mass Spectrometry, Proteomics More...
mol wt   monoisotopic mol wt 1622.90 Da
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... GAPDH(2597)
pig ... GAPDH(396823)

Description

Application

Protein-AQUA is a powerful, enabling technology that facilitates focused, quantitative studies of not only specific protein expression, but specific amino acid modification as well, using stable-isotope–labeled peptides and mass spectrometry.

Legal Information

This method was developed by Dr. Steve Gygi and colleagues at Harvard Medical School.1 Limited use of this method is permitted under a licensing arrangement with Harvard Medical School.

AQUA is a trademark of Harvard Medical School

Protein AQUA is a trademark of Harvard Medical School

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?