Synonym: Anti-Cleavage stimulation factor, 3′ pre-RNA, subunit 3, 77 kDa
| Related Categories | Alphabetical Index, Antibodies, Antibodies for RNA Regulation and Translation, Antibodies to RNA Binding Proteins, Antibodies to Ribosomal Proteins and Translation Factors, |
| species reactivity | canine, zebrafish, human, mouse, rat, bovine, chicken, |
| application(s) | western blot: suitable |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | lyophilized powder |
| immunogen sequence |
IEAEIKAKNYDKVEKLFQRCLMKVL |
| storage temp. | −20°C |
| Gene Information | human ... CSTF3(1479) |
| NCBI accession no. |
NP_001317 |
synthetic peptide corresponding to a region of human CSTF3 with an internal ID of P10391
Lyophilized from PBS buffer with 2% sucrose
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?