Synonym: Anti-Collapsin response mediator protein 1, Anti-DPYSL1, Anti-DRP-1, Anti-DRP1
| Related Categories | Alphabetical Index, Antibodies, Antibodies for Neurobiology, Antibodies to Neural Proteins, CR-CR, |
| species reactivity | zebrafish, bovine, canine, chicken, human, mouse, rat |
| application(s) | western blot: suitable |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | lyophilized powder |
| immunogen sequence |
PSKHQPPPIRNLHQSNFSLSGAQID |
| storage temp. | −20°C |
| Gene Information | human ... CRMP1(1400) |
| NCBI accession no. |
NP_001304 |
synthetic peptide corresponding to a region of human CRMP1 with an internal ID of P12528
Lyophilized from PBS buffer with 2% sucrose
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?