CAS Number 2134-24-9
Empirical Formula (Hill Notation) C26H21NO3
Molecular Weight 395.45
EC Number 218-364-6
MDL number MFCD00021607
PubChem Substance ID 24891646
| Related Categories | A - H, Amino Acids, Biochemicals and Reagents, Modified Amino Acids |
| storage temp. | −20°C |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?