MDL number MFCD02096040
Popular Documents: Datasheet (PDF)
| Related Categories | Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Cycle, Antibodies in Array, |
| species reactivity | human |
| application(s) | microarray: suitable |
| western blot: 1:1,000 using an extract from a cultured human chronic myelogenous leukemia cell line, K562 | |
| clone | DCS-193, monoclonal |
| antibody form | ascites fluid |
| isotype | IgG1 |
| mol wt | antigen mol wt 65 kDa |
| contains | 15 mM sodium azide |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | human ... CDC25C(995) |
recombinant human Cdc25c.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?