Synonym: Cysteine protease inhibitor
CAS Number 81989-95-9
MDL number MFCD00130887
| Related Categories | A to C, Application Index, Bioactive Small Molecules, Biochemicals and Reagents, CI-CY, |
| form | buffered aqueous glycerol solution |
| potency | 40-80 BAEE units per mg protein 50% inhibition of papain (pH 6.8 at 40 °C) |
| storage temp. | −20°C |
Protein determined by E
Solution in 10 mM Tris buffer, pH 8.0, containing 50% glycerol.
Inhibitor of cysteine proteases, including papain and dipeptidyl peptidase III. Effective concentration equimolar with protease. A docking model has been proposed for the mechanism of inhibition of papain.1
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?