C4523 Sigma

N-Z-Case® TT

Moderately digested casein enzymatic hydrolysate, from bovine milk

DOWNLOAD MSDS (PDF)

Synonym: Peptone from casein

Properties

Related Categories Media Components, Microbial Media for Molecular Biology, Molecular Biology, Protein Hydrolysates
biological source   from bovine milk
nitrogen analysis   ≥5% amino, ≥12% total

Description

Application

Originally formulated for use in media for the production of tetanus toxin.

Reconstitution

Recommended concentration: 25 g/L

Legal Information

N-Z-Case is a registered trademark of Kerry Group

Price and Availability

Customers Also Viewed

for microbiology

Casein enzymatic hydrolysate, from bovine milk

from bovine milk, Casein enzymatic hydrolysate

for microbiology

Casein enzymatic hydrolysate, from bovine milk

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?