Synonym: autoradiography cassettes, xray film cassettes
| Related Categories | Autoradiography, Autoradiography Exposure Cassettes, Carestream Kodak, Carestream Kodak Autoradiography, Electrophoresis Supplies, |
Lightweight, durable, vinyl-covered aluminum cassettes, light-tight with single locking clip.
When used at ultra-cold temperature, allow to come to room temperature before opening to avoid cracking polyurethane hinge.
BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company
Customers Also Viewed
pack of 1 gal
For 14 in. x 17 in. (35 x 43 cm) film
for 35 × 43 cm film
for 8 × 10 in. film