Synonym: Aminoacylproline Carboxypeptidase, CPP, Penicillocarboxypeptidase S-1, Proline Carboxypeptidase
CAS Number 9075-64-3
MDL number MFCD00130721
| Related Categories | 3.4.x.x Peptidases, 3.x.x.x Hydrolases, Application Index, Biochemicals and Reagents, Carboxypeptidase P, |
| form | lyophilized powder |
| packaging | vial of ≥100 units |
| storage temp. | −20°C |
One unit will hydrolyze 1.0 μmole of N-CBZ-Glu-Tyr to N-CBZ-
Lyophilized powder containing sodium citrate
Customers Also Viewed
(Type II-PMSF treated), ≥50 units/mg protein, aqueous suspension
lyophilized powder, ≥3 units/mg protein
≥50 units/mg protein
lyophilized powder, ≥50 units/mg protein
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?