MDL number MFCD06798712
Popular Documents: Datasheet (PDF)
| Related Categories | Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Embryonic Stem Cells, Antibodies for Neural Stem Cells, |
| species reactivity | mouse, monkey, rat, human |
| application(s) | immunocytochemistry: suitable |
| indirect ELISA: suitable | |
| microarray: suitable | |
| western blot: 4-8 μg/mL using whole extract of cultured human 293T cells treated with MG-132 | |
| clone | BCS-45, monoclonal |
| antibody form | purified immunoglobulin |
| form | buffered aqueous solution |
| isotype | IgG1 |
| mol wt | antigen mol wt ~94 kDa |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information |
human ... CTNNB1(1499) mouse ... Ctnnb1(12387) rat ... Ctnnb1(84353) |
synthetic phosphopeptide corresponding to the region that contains serine 45 of human β-catenin.
Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?