CLS3298 Sigma

Corning® CellBIND® Surface cell culture flasks

CellBIND 175cm2 rectangular angled neck cell culture flask w/ phenolic-style cap, PS, sterile, 50/cs

DOWNLOAD MSDS (PDF)

Synonym: cell culture flasks, tissue culture flasks

Properties

Related Categories Cell Culture Supplies, Cell Culture Surfaces, CellBind Surface, Corning, Corning Cell Culture Flasks,
material   angled neck
  cap (phenolic-style)
  flask rectangular
  high-density polyethylene cap
  polystyrene rectangular
sterility   sterile; γ-irradiated
feature   graduations
packaging   case of 50
  pack of 5
mfr. no.   Corning, 3298
maximum volume   250 mL
surface area   175 cm2
working volume   35-52.5 mL
matrix active group   CellBIND® surface treatment
suitability   suitable for (cell culture applications; optimal for adherent cell culturing)

Description

General description

The Corning CellBIND surface is produced by a microwave process which makes a more hydrophilic surface giving more consistent, even cell attachment, increased cell growth and yields. CellBIND enhances cell attachment and growth under difficult conditions, including reduced serum or serum-free medium.

CellBIND may provide a more ecominic alternative to expensive and unstable biological coatings.

Both enzymatic and non-enzymatic dissociating solutions have been used to remove and harvest cells from CellBIND surfaces, scraping is not recommended due to the increased strength of cell attachment.

• Manufactured from optically clear virgin polystyrene.
• Treated CellBIND surface treatment for enhanced cell attachment
• 100% integrity tested.
• Sterilized by gamma irradiation and certified nonpyrogenic

Legal Information

CellBIND is a registered trademark of Corning, Inc.

Corning is a registered trademark of Corning, Inc.

Price and Availability

Customers Also Viewed

Corning<SUP>®</SUP> CellBIND<SUP>®</SUP> Surface cell culture flasks

CellBIND 25cm2 rectangular canted neck cell culture flask w/ Vent Cap, PS, sterile, 200/cs

Corning<SUP>®</SUP> CellBIND<SUP>®</SUP> Surface cell culture flasks

CellBIND 75cm2 rectangular canted neck cell culture flask w/ Vent Cap, PS, sterile, 100/cs

Corning<SUP>®</SUP> CellBIND<SUP>®</SUP> Surface cell culture flasks

CellBIND 150cm2 rectangular canted neck cell culture flask w/ Vent Cap, PS, sterile, 50/cs

Corning<SUP>®</SUP> CellBIND<SUP>®</SUP> Surface cell culture flasks

CellBIND 175cm2 angled neck cell culture flask w/ Vent Cap, PS, sterile, 50/cs

Corning<SUP>®</SUP> CellBIND<SUP>®</SUP> Surface cell culture flasks

CellBIND 175cm2 angled neck cell culture flask w/ Vent Cap and Bar Code, PS, sterile, 84/cs

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?